Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talc, Powder, USP, Spectrum™ Chemical
SDP

Catalog No. 18607198
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

T1005, 14807-96-6, 3MgO·4SiO2·H2O

Spectrum™ Chemical Talc, Powder, USP is used as a carrier for pharmaceuticals with a disinfectant, astringent, anti-itching or cooling effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 14807-96-6
Molecular Formula H2Mg3O12Si4
Molecular Weight (g/mol) 379.26
MDL Number MFCD00792903
InChI Key KNXVOGGZOFOROK-UHFFFAOYSA-N
IUPAC Name trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
SMILES [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Grade USP
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.