Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TALDO1 Monoclonal Antibody (OTI1E5)
GREENER_CHOICE

Catalog No. PIMA527221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527221 Supplier Invitrogen™ Supplier No. MA527221
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TALDO1 Monoclonal Antibody for Western Blot, IHC (P)

Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TALDO1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TALDO1
Gene Accession No. P37837
Gene Alias dihydroxyacetone transferase; glycerone transferase; TAL; TALDO; TALDO1; TALDOR; TALH; TAL-H; testicular secretory protein Li 56; transaldolase; transaldolase 1
Gene Symbols TALDO1
Host Species Mouse
Immunogen Full length human recombinant protein of TALDO1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6888
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.