Learn More
TALDO1 Mouse anti-Human, Clone: OTI4A7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809920

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.Specifications
TALDO1 | |
Monoclonal | |
Unconjugated | |
TALDO1 | |
TAL, TAL-H, TALDOR, TALH | |
Mouse | |
Affinity Chromatography | |
RUO | |
6888 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P37837 | |
TALDO1 | |
Full length human recombit protein of human TALDO1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.