Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TALDO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565496 Supplier Invitrogen™ Supplier No. PA565496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVGRILDWHVA NTDKKSYEPL EDPGVKSVTK IYNYYKKFSY KTIVMGASFR NTGEIKALAG CDFLTISPKL LGEL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TALDO1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TALDO1
Gene Accession No. P37837
Gene Alias dihydroxyacetone transferase; glycerone transferase; TAL; TALDO; TALDO1; TALDOR; TALH; TAL-H; testicular secretory protein Li 56; transaldolase; transaldolase 1
Gene Symbols TALDO1
Host Species Rabbit
Immunogen Recombinant Human TALDO1. Recombinant protein control fragment (Product #RP-104846).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.