Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Talin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550838
Description
The antibody detects endogenous levels of total Talin protein.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
Specifications
Talin 1 | |
Polyclonal | |
Unconjugated | |
TLN1 | |
ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21894, 7094 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P26039, Q9Y490 | |
TLN1 | |
Synthetic peptide corresponding to residues near the N terminal of human talin 1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction