Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Talin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582162 Supplier Invitrogen™ Supplier No. PA582162
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLYTAKEAGG NPKQAAHTQE ALEEAVQMMT EAVEDLTTTL NEAASAAGVV GGMVDSITQA INQLDEGPMG EPEGSFVDYQ TTMVRTAKAI AVTVQEMVTK SNTSP Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 99%.

Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Talin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TLN1
Gene Accession No. Q9Y490
Gene Alias ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1
Gene Symbols TLN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TLN1. Recombinant protein control fragment (Product #RP-88634).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7094
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.