Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Talin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584219
Description
Immunogen sequence: IEAGKLVDRS VENCVRACQA ATTDSELLKQ VSAAASVVSQ ALHDLLQHVR QFASRG.
This gene encodes a protein related to talin 1, a cytoskeletal protein that plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. This protein has a different pattern of expression compared to talin 1 but, like talin 1, is thought to associate with unique transmembrane receptors to form novel linkages between extracellular matrices and the actin cytoskeleton.
Specifications
Talin 2 | |
Polyclonal | |
Unconjugated | |
Tln2 | |
5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; mKIAA0320; RGD1565416; talin 2; talin-2; TLN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4G6 | |
Tln2 | |
Recombinant protein corresponding to Human TLN2. Recombinant protein control fragment (Product #RP-108207). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction