Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin1 Antibody (93E12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250319H
Description
Talin1 Monoclonal specifically detects Talin1 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Talin1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
TLN1 | |
Recombinant protein fragment | |
0.1 mL | |
Cytoskeleton Markers | |
7094 | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
93000000000000 | |
Western Blot, ELISA, Immunohistochemistry | |
ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross react with Talin2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction