Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin1 Antibody (TA205), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP121642PCP
Description
Talin1 Monoclonal specifically detects Talin1 in Human, Chicken, Rabbit samples. It is validated for Flow Cytometry.Specifications
Talin1 | |
Monoclonal | |
PerCP | |
Q9Y490 | |
TLN1 | |
Human platelet talin. | |
0.1 mL | |
Cytoskeleton Markers | |
7094 | |
Human, Chicken, Rabbit | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
TA205 | |
Flow Cytometry | |
ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Immunoblots the 225 kDa talin band on SDS gels of platelets and other cells. It may be used to stain adhesion plaques in fixed and permeabilized fibroblasts. TA205 recognizes an epitope within residues 139-433. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction