Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin2 Antibody (68E7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250322AF405
Description
Talin2 Monoclonal specifically detects Talin2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Talin2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TLN2 | |
recombinant protein fragment | |
0.1 mL | |
Cytoskeleton Markers | |
83660 | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
680000000 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp451B1011, DKFZp686I0976, DKFZp686K0979, ILWEQ, KIAA0320, talin2, talin-2, TLN 2, TLN2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross react with Talin1. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction