Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin2 Antibody (SR1434), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32163650UL
Description
Talin2 Monoclonal antibody specifically detects Talin2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Talin2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1434 | |
Western Blot 1:500-1:2000 | |
DKFZp451B1011, DKFZp686I0976, DKFZp686K0979, ILWEQ, KIAA0320, talin2, talin-2, TLN 2, TLN2 | |
A synthesized peptide derived from human Talin2 (Uniprot #: Q9Y4G6) | |
50 μg | |
Cell Biology, Cellular Markers | |
83660 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction