Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle Talon™ Drive-In and Drive-Out
GREENER_CHOICE

Catalog No. 17989777
Click to view available options
Weight (English):
120 lb.
142 lb.
198 lb.
252 lb.
293 lb.
475 lb.

Drive-In Drive-Out Berm allows the user to drive in and out without having to lower the end walls. Fuel and chemical resistant.

  • Ideal for fuel use
  • Constructed of XR5 Fabric
  • Easy to assemble
  • Lap joint welds for maximum strength
  • Designed for quick deployment
  • Made in the USA

Specifications

Material XR-5
Type Spill Containment Pallet
Weight Capacity Heavyweight
Weight (English) 142 lb.
Weight (Metric) 64.4 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.