Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva TALON™ Superflow Medium

Catalog No. 45002390 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-390 50 mL
45-002-389 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-390 Supplier Cytiva Supplier No. 28957502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offers different selectivity compared to nickel-charged media. Cytiva TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.

Features:

  • Ligand - TALON Superflow Cobalt-IMAC
  • Matrix - Cross-linked agarose, 6%
  • Particle Size - 60 -160μm
  • Binding Capacity/mL - Up to 20 mg histidine-tagged protein
  • pH stability working range - 3 to 12
  • pH stability Cleaning-in-Place (CIP) - 2 - 14 Co2+stripped medium
  • Complete packsize - 10mL

 

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
For Use With (Application) For PurifiCatIon of Histidine-Tagged Recombinant Pby Immobilized Metal Affinity Chromatography
Storage Requirements 4°C to 8°C
Binding Capacity Up to 20 mg Histidine-tagged Protein
Product Type Talon Superflow Media
Particle Size 60 to 160 μm
Matrix 6% Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Talon Superflow Cobalt-IMAC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.