Learn More
Invitrogen™ Talquetamab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Supplier:  Invitrogen™ MA541863
Catalog No. PIMA541863
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Talquetamab (JNJ-64407564) is a bispecific antibody that targets CD3, which is expressed on T-cells, and GPRC5D, which is highly expressed in multiple myeloma cells. Talquetamab (JNJ-64407564) is being investigated in a Phase 1 clinical study to treat relapsed or refractory multiple myeloma.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Talquetamab Humanized
Recombinant Monoclonal
Unconjugated
JNJ-64407564
Human CD3E/GPRC5D.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
1.75 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A/G
RUO
Human
Antibody
IgG4 κ, IgG4 λ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.