Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tamibarotene, PROMO, Thermo Scientific Chemicals

Catalog No. J64028MAPM
Click to view available options
Quantity:
10 mg

Applications

Tamibarotene is a retinoic acid receptor α (RARα) agonist that induces differentiation (ED50 = 0.79 nM) and apoptosis of HL-60 cells in vitro. Exhibits antiproliferative effects against a variety of human tumor cells lines (mean values of 35, 40 and 60% growth inhibition at 0.1, 1 and 10 μM respectively).

Solubility

Soluble to 50 mM in DMSO

Notes

Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94497-51-5
Molecular Formula C22H25NO3
Molecular Weight (g/mol) 351.45 g/mol
MDL Number MFCD00866188
InChI Key MUTNCGKQJGXKEM-UHFFFAOYSA-N
Synonym tamibarotene, retinobenzoic acid, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl carbamoyl benzoic acid, amnolake, tamibaro, amnoid, unii-08v52gz3h9, 4-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl carbamoyl benzoic acid, chembl25202
PubChem CID 108143
ChEBI CHEBI:32181
IUPAC Name 4-[(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)carbamoyl]benzoic acid
SMILES CC1(C)CCC(C)(C)C2=CC(NC(=O)C3=CC=C(C=C3)C(O)=O)=CC=C12

Specifications

Color White
Quantity 10 mg
Merck Index 14,9047
Solubility Information Soluble in DMSO
Physical Form Solid
Chemical Name or Material Tamibarotene
RTECSNumber DH6940000
TSCA No

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.