Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAMRA Antibody (5G5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222472
Description
TAMRA Monoclonal specifically detects TAMRA in Non-species specific samples. It is validated for Western Blot, Immunoprecipitation.Specifications
TAMRA | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Immunoprecipitation 10 ug | |
Mouse | |
Protein G purified | |
RUO | |
This antibody reacts with Chemical. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunoprecipitation | |
5G5 | |
Unconjugated | |
BP-25606, F07-0040, TAMRA Azide, TAMRA Azide, isomer 5, tetramethylrhodamine azide | |
TAMRA molecule | |
100 μg | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction