Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TANC1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP152630C
Description
TANC1 Polyclonal antibody specifically detects TANC1 in Human samples. It is validated for Western BlotSpecifications
TANC1 | |
Polyclonal | |
Western Blot | |
FLJ13360, FLJ41166, KIAA1728, protein TANC1, rolling pebbles homolog B, ROLSB, TANC, tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1, tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1, TPR domain, ankyrin-repeat and coiled-coil-containing | |
The immunogen for this product maps to a region between residue 1 and 50 of human Tetratricopeptide Repeat, Ankyrin Repeat and Coiled-Coil Containing 1 using the numbering given in entry NP_203752.2 (GeneID 85461). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
85461 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction