Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TANC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557797 Supplier Invitrogen™ Supplier No. PA557797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQEGLQSKGR PVSPQSRAGI GKSLREPVAQ PGLLLQPSKQ AQIVKTSQHL GSGQSAVRNG SMKVQISSQN PPPSPMPGRI AATP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.

TANC1 (Tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1) is a PSD-95-interacting synaptic protein that is proposed to play a role in the regulation of dendritic spines and excitatory synapses and spatial memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TANC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TANC1
Gene Accession No. Q9C0D5
Gene Alias 1200003E16Rik; KIAA1728; mKIAA1728; Protein TANC1; rolling pebbles homolog B; ROLSB; Tanc; TANC1; tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1; tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1; TPR domain, ankyrin-repeat and coiled-coil domain-containing protein 1; TPR domain, ankyrin-repeat and coiled-coil-containing; TPR domain, ankyrin-repeat and coiled-coil-containing protein
Gene Symbols TANC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TANC1. Recombinant protein control fragment (Product #RP-96006).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85461
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.