Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TANC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981051
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1051 Supplier Bioss Supplier No. BS11037R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TANC1 (Tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1) is a PSD-95-interacting synaptic protein that is proposed to play a role in the regulation of dendritic spines and excitatory synapses and spatial memory.

Specifications

Antigen TANC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TANC1
Gene Accession No. Q9C0D5
Gene Alias KIAA1728; Protein TANC1; rolling pebbles homolog B; ROLSB; TANC; TANC1; tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1; Tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1; TPR domain, ankyrin-repeat and coiled-coil-containing
Gene Symbols TANC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TANC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85461
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.