Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Tangential Flow Filtration (TFF)- MV Filter
SDP

Catalog No. NB452216 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB452216 1 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB452216 Supplier Novus Biologicals™ Supplier No. NBP326836
Add to Cart
Add to Cart

Filter cartridge of hollow polysulfone fibers used for separating different EVs by size

Specifications

Content And Storage Store at room temperature.
Quantity 1 Units
Sample Type Diluted matrices (urine and cell culture supernatant)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.