Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TANGO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551818 Supplier Invitrogen™ Supplier No. PA551818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPVNVQRRED SATEGSHRLI LAANRDEFYS RPSKLADFWG NNNEILSGLD MEEGKEGGTW LGISTRGKLA ALTNYLQPQL DWQARGRGEL VTHFLTTDVD SLSYLKKVSM EGHLYNGFNL IAADLSTAKG DVICYYGNRG EPDPIVLT Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

This gene belongs to the transport and Golgi organization family, whose members are predicted to play roles in secretory protein loading in the endoplasmic reticulum. Depletion of this gene in Drosophila S2 cells causes fusion of the Golgi with the ER. In mouse tissue culture cells, this protein co-localizes with a mitochondrially targeted mCherry protein and displays very low levels of co-localization with Golgi and peroxisomes. Allelic variants of this gene are associated with rhabdomyolysis, metabolic crises with encephalopathy, and cardiac arrhythmia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TANGO2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TANGO2
Gene Accession No. Q6ICL3
Gene Alias C17H22orf25; C22orf25; C26H22orf25; D16H22S680E; DKFZP761P1121 protein; hypothetical protein DKFZp761P1121; MECRCN; RGD1310348; ser/Thr-rich protein T10 in DGCR region; T10; TANGO2; transport and golgi organization 2; transport and golgi organization 2 homolog; transport and golgi organization 2 homolog (Drosophila); transport and Golgi organization protein 2 homolog; Uncharacterized protein C22orf25; uncharacterized protein C22orf25 homolog; wu:fj48d05; zgc:100952
Gene Symbols TANGO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TANGO2. Recombinant protein control fragment (Product #RP-91525).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128989
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.