Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tankyrase 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220559
Description
Tankyrase 1 Polyclonal specifically detects Tankyrase 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Tankyrase 1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
EC 2.4.2.30, PARP-5a, PARP5APARPL, pART5, Poly [ADP-ribose] polymerase 5A, Tankyrase I, tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase, tankyrase-1, TIN1TANK1, TINF1TNKS-1, TNKS1TRF1-interacting ankyrin-related ADP-ribose polymerase | |
Rabbit | |
142 kDa | |
0.1 mL | |
Apoptosis, Chromatin Research, DNA Repair | |
8658 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O95271 | |
TNKS | |
Recombinant protein encompassing a sequence within the N-terminus region of human Tankyrase. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction