Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tankyrase-1/TNKS Mouse anti-Human, Mouse, Unconjugated, Clone: 7D1.2, MilliporeSigma™

Catalog No. MABC286 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC286 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC286 Supplier MilliporeSigma Supplier No. MABC286
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tankyrase-1/TNKS Antibody, clone 7D1.2 is validated for use in Western Blotting and Immunocytochemistry and Immunohistochemistry for the detection of Tankyrase-1/TNKS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tankyrase-1/TNKS
Applications Western Blot, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 7D1.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Tankyrase-1;TANK1;ADP-ribosyltransferase diphtheria toxin-like 5;ARTD5;Poly [ADP-ribose] polymerase 5A;TNKS-1;TRF1-interacting ankyrin-related ADP-ribose polymerase;Tankyrase I
Gene Symbols TNKS;PARP5A;PARPL;TIN1;TINF1;TNKS1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to Human Tankyrase-1/TNKS.
Molecular Weight of Antigen ∽142 kDa observed.Uncharacterized bands may be observed in some lysates.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_003738
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.