Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tankyrase 2 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5121562
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121562 100 μL
PIPA5121563 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5121562 Supplier Invitrogen™ Supplier No. PA5121562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Poly-ADP-ribosyltransferase involved in various processes such as Wnt signaling pathway, telomere length and vesicle trafficking (PubMed:11739745, PubMed:11802774, PubMed:19759537, PubMed:21478859, PubMed:23622245, PubMed:25043379). Acts as an activator of the Wnt signaling pathway by mediating poly-ADP-ribosylation of AXIN1 and AXIN2, 2 key components of the beta-catenin destruction complex: poly-ADP-ribosylated target proteins are recognized by RNF146, which mediates their ubiquitination and subsequent degradation (PubMed:19759537, PubMed:21478859). Also mediates poly-ADP-ribosylation of BLZF1 and CASC3, followed by recruitment of RNF146 and subsequent ubiquitination (PubMed:21478859). Mediates poly-ADP-ribosylation of TERF1, thereby contributing to the regulation of telomere length (PubMed:11739745). Stimulates 26S proteasome activity (PubMed:23622245). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tankyrase 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TNKS2
Gene Accession No. Q9H2K2
Gene Alias 5430432P15Rik; AA517131; ADP-ribosyltransferase diphtheria toxin-like 6; AI662480; ARTD6; PARP5B; PARP-5b; PARP5C; PARP-5c; pART6; Poly [ADP-ribose] polymerase 5B; Poly [ADP-ribose] polymerase tankyrase-2; Protein poly-ADP-ribosyltransferase tankyrase-2; TANK2; tankyrase 2; tankyrase II; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2; tankyrase-2; Tankyrase-like protein; tankyrase-related protein; TNKL; tnks 2; TNKS2; TNKS-2; TRF1-interacting ankyrin-related ADP-ribose polymerase 2
Gene Symbols TNKS2
Host Species Rabbit
Immunogen Ser959-Gly1166 (Accession Q9H2K2), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.