Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tankyrase Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA517931
Description
Tankyrase Polyclonal Antibody for Western Blot
Poly-ADP-ribosyltransferase involved in various processes such as Wnt signaling pathway, telomere length and vesicle trafficking (PubMed:10988299, PubMed:11739745, PubMed:16076287, PubMed:19759537, PubMed:21478859, PubMed:22864114, PubMed:23622245, PubMed:25043379). Acts as an activator of the Wnt signaling pathway by mediating poly-ADP-ribosylation (PARsylation) of AXIN1 and AXIN2, 2 key components of the beta-catenin destruction complex: poly-ADP-ribosylated target proteins are recognized by RNF146, which mediates their ubiquitination and subsequent degradation (PubMed:19759537, PubMed:21478859). Also mediates PARsylation of BLZF1 and CASC3, followed by recruitment of RNF146 and subsequent ubiquitination (PubMed:21478859). Mediates PARsylation of TERF1, thereby contributing to the regulation of telomere length (PubMed:11739745). Involved in centrosome maturation during prometaphase by mediating PARsylation of HEPACAM2/MIKI (PubMed:22864114). May also regulate vesicle trafficking and modulate the subcellular distribution of SLC2A4/GLUT4-vesicles (PubMed:10988299). May be involved in spindle pole assembly through PARsylation of NUMA1 (PubMed:16076287). Stimulates 26S proteasome activity (PubMed:23622245). [UniProt].
Specifications
| Tankyrase | |
| Polyclonal | |
| Unconjugated | |
| TNKS | |
| 4930554K12Rik; ADP-ribosyltransferase diphtheria toxin-like 5; AI662855; ARTD5; C86528; D130072O21Rik; HGNC:11941; LOW QUALITY PROTEIN: tankyrase-1; mTNKS1; PARP5A; PARP-5a; PARPL; pART5; Poly [ADP-ribose] polymerase 5A; Poly [ADP-ribose] polymerase tankyrase-1; poly [ADP-ribose] polymerase tankyrase-1; tankyrase-1; Protein poly-ADP-ribosyltransferase tankyrase-1; TANK1; tankyrase; tankyrase 1; tankyrase I; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase; tankyrase-1; TIN1; TINF1; TNKS; TNKS1; TNKS-1; TRF1-interacting ankyrin-related ADP-ribose polymerase; TRF1-interacting ankyrin-related ADP-ribose polymerase 1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 8658 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O95271 | |
| TNKS | |
| Synthetic peptide sequence (RECOMBINANT) corresponding to the N-terminus amino acids of TNKS. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction