Learn More
Tantalum(V) bromide, 99.9% (metals basis)
Description
The Tantalum Bromide Cluster is used for the preparation of heavy-atom derivatives for structure determination of biological macromolecules by X-ray analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 348.8°C |
| Melting Point | 265°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN3260 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 580.49 |
| Percent Purity | 99.9% |
| Odor | Pungent |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 236-618-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.