Learn More
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis)
Description
Tantalum(V) chloride is used to form mixed chloro-aryloxide compounds, such as [Ta(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. It is also used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 242°C |
Melting Point | 216°C |
Physical Form | Resublimed Crystalline |
Quantity | 5 g |
Assay Percent Range | 99.99%, metal basis |
Solubility Information | Soluble in water (reacts). Also soluble in ethanol,ether and carbon tetrachloride. |
Formula Weight | 358.21 |
Sensitivity | Moisture sensitive |
Density | 3.68 |
Chemical Name or Material | Tantalum(V) Chloride, Ampouled under argon |
Safety and Handling
Hazard Category | H302-H314-H335 |
Hazard Statement | When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | DOT Class: 8; Packaging Group: II |
EINECSNumber | 231-755-6 |
RTECSNumber | WW5600000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.