Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum(V) oxide, 99% (metal basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1470909

Quantity 10g

 View more versions of this product

Catalog No. AA1470909


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1314-61-0
99% (metals basis)
441.89
Tantalum pentoxide
Powder
Ta2O5
MFCD00011254
Tantalum(V) oxide

Specifications

1314-61-0
99% (metals basis)
441.89
Tantalum pentoxide
Tantalum(V) oxide
Powder
Ta2O5
MFCD00011254
10g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 215-238-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit