Learn More
GSC Go Science Crazy Tantalus Cup
SCI_ED
Catalog No. s04817 Shop All GSC Products
Encompass
Click to view available options
:
Tantalus Cup
A self-siphoning cup that can never be filled. The Tantalus Cup helps demonstrate Pascal's principle, or how a siphon works.
  • A glass cup with a U-tube attached
  • When water reaches the top of the inverted U-tube, a siphon action begins which drains the water until the end of the U-tube is exposed
  • When the cup is filled with water, the water is forced up the inverted U-tube so that equilibrium is maintained in the cup
  • As the water level reaches the top of the U-tube it is begins to fall through the siphon
  • As the water falls, it creates a vacuum which causes more water to be forced into the tube
Specifications
Product Type Tantalus Cup
Material Glass
WARNING: Cancer - www.P65Warnings.ca.gov
Not a toy. Product intended for ages 13 and up. May contain latex!
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.