Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAO2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220562 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220562 Supplier Novus Biologicals Supplier No. NBP220562
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAO2 Polyclonal specifically detects TAO2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TAO2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UL54
Gene Alias EC 2.7.11, KIAA0881PSK-1, Kinase from chicken homolog C, MAP3K17PSK1-BETA, Prostate-derived STE20-like kinase 1, PSK1EC 2.7.11.1, PSKhKFC-C, serine/threonine-protein kinase TAO2, TAO kinase 2, TAO1, TAO2prostate derived STE20-like kinase PSK, Thousand and one amino acid protein 2, thousand and one amino acid protein kinase
Gene Symbols TAOK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TAOK2. The exact sequence is proprietary.
Molecular Weight of Antigen 138 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9344
Test Specificity Mouse. Species reactivity based on sequence identity: Human (100%),Bovine (100%).
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.