Learn More
Promega TAOK1 Kinase Enzyme System, 10microg
Catalog No. PRV4090 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4090 10 μg
1 options
Catalog No. PRV4090 Supplier Promega Supplier No. V4090
TAOK1 is a serine/threonine-protein kinase involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs).
  • Easily Screen and Profile TAOK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C