Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAOK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA124577
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA124577 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA124577 Supplier Invitrogen™ Supplier No. PA124577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-24577 detects Serine/threonine kinase TAO kinase 1 in Human samples.

TAOK1 is a serine/threonine-protein kinase involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs). The activation of and binding to MEK3 by TAOK1 implicates TAOK1 in the regulation of the p38-containing stress-responsive MAP kinase pathway. A microtubule affinity-regulating kinase, TAOK1 (also known as MARKK) is an important regulator of mitotic progression, required for both chromosome congression and checkpoint-induced anaphase delay. TAOK1 is involved in various processes such as p38/MAPK14 stress-activated MAPK cascase, DNA damage response, and regulations of cytoskeleton stability. It phosphorylates MAP2K3, MAP2K6, and MARK2. TAOK1 acts as an activator of the p38/MAPK cascade by mediating phosphorylation and subsequent activation of the upsteam MAP2K3 and MAP2K6 kinases. It is involved in G-protein couples receptor signaling to p38/MAPK14. In response to DNA damage, TAOK1, is involved in the G2/M transition DNA damage checkpoint by activating the p38/MAPK14 stress-activated MAPK cascade by phosphorylaton of MAP2K3 and MAP2K6. In addition, it acts as a regulator of cytoskeleton stability and a regulator of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAOK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TAOK1
Gene Accession No. O88664, Q5F2E8, Q7L7X3
Gene Alias 2810468K05Rik; AU020252; D130018F14Rik; FLJ14314; h hTAOK1; H KIAA1361; hKFC-B; hTAOK1; KFC-B; Kiaa1361; Kinase from chicken homolog B; MAP3K16; MARK Kinase; Markk; microtubule affinity regulating kinase kinase; mKIAA1361; prostate-derived STE20-like kinase 2; prostate-derived sterile 20-like kinase 2; PSK2; PSK-2; serine/threonine protein kinase TAO1; serine/threonine protein kinase TAO1 homolog; serine/threonine-protein kinase TAO1; STE20-like kinase PSK2; tao; TAO kinase 1; TAO1; TAOK1; Thousand and one amino acid protein 1; thousand and one amino acid protein kinase 1
Gene Symbols TAOK1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human TAOK1.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100059705, 100342573, 100622172, 100758772, 216965, 286993, 480632, 536532, 57551
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Frog, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.