Learn More
TAOK1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300524A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
TAOK1 is a TAO type protein kinase. TAOK1 expression has been documented in rat brain. ESTs have been isolated from human normal adrenal, bone and breast and cancerous duodenum, cervix and germ cell.Specifications
TAOK1 | |
Polyclonal | |
Unconjugated | |
TAOK1 | |
FLJ14314, hKFC-B, hTAOK1, KFC-B, KIAA1361, kinase from chicken homolog B, MAP3K16, MARK Kinase, MARKK, microtubule affinity regulating kinase kinase, prostate-derived STE20-like kinase 2, prostate-derived sterile 20-like kinase 2, PSK2, PSK-2, serine/threonine protein kinase TAO1 homolog, STE20-like kinase PSK2, TAO1, TAOK1, thousand and one amino acid protein 1, thousand and one amino acid protein kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57551 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q7L7X3 | |
TAOK1 | |
Between 400 and 450 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.