Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Taok2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529248 Supplier Invitrogen™ Supplier No. PA529248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (95%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TAOK2 (TAO1) is a serine/threonine kinase that may be an important regulator of the stress-responsive p38 MAP kinase pathway. In vitro, TAO (thousand and one amino acid) protein kinase 2 (TAO2) activates MAP/ERK kinases (MEK ) 3, 4, and 6 toward their substrates p38 MAP kinase JNK/SAPK (Chen et al., 1999; Chen and Cobb, 2001). This and more recent work has led to the proposal that the TAO protein kinases play an essential role in signaling from physiological agonist to the stress -responsive p38 MAPK (Chen et al., 2003). Autophosphorylation of TAO may play a role in the mechanism of TAO activation. The MEK binding domain of TAO is autophosphorylated on both serine and threonine residues and Ser181 is located within this domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Taok2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TAOK2
Gene Accession No. Q6ZQ29, Q9UL54
Gene Alias 1110033K02Rik; B230344N16; hKFC-C; Kiaa0881; Kinase from chicken homolog C; MAP3K17; mKIAA0881; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived sterile 20-like kinase 1; PSK; PSK1; PSK-1; PSK1-BETA; serine/threonine protein kinase TAO2; serine/threonine-protein kinase TAO2; serine-threonine kinase; tao; TAO kinase 2; TAO kinase 2TAO kinase 2; TAO1; TAO2; TAOK2; Thousand and one amino acid protein 2; thousand and one amino acid protein kinase 2; UNQ2971/PRO7431
Gene Symbols TAOK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TAOK2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381921, 9344
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.