Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAOK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567854 Supplier Invitrogen™ Supplier No. PA567854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

TAOK2 (TAO1) is a serine/threonine kinase that may be an important regulator of the stress-responsive p38 MAP kinase pathway. In vitro, TAO (thousand and one amino acid) protein kinase 2 (TAO2) activates MAP/ERK kinases (MEK ) 3, 4, and 6 toward their substrates p38 MAP kinase JNK/SAPK (Chen et al., 1999; Chen and Cobb, 2001). This and more recent work has led to the proposal that the TAO protein kinases play an essential role in signaling from physiological agonist to the stress -responsive p38 MAPK (Chen et al., 2003). Autophosphorylation of TAO may play a role in the mechanism of TAO activation. The MEK binding domain of TAO is autophosphorylated on both serine and threonine residues and Ser181 is located within this domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAOK2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAOK2
Gene Accession No. Q6ZQ29, Q9JLS3, Q9UL54
Gene Alias 1110033K02Rik; B230344N16; hKFC-C; Kiaa0881; Kinase from chicken homolog C; MAP3K17; mKIAA0881; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived sterile 20-like kinase 1; PSK; PSK1; PSK-1; PSK1-BETA; serine/threonine protein kinase TAO2; serine/threonine-protein kinase TAO2; serine-threonine kinase; tao; TAO kinase 2; TAO kinase 2TAO kinase 2; TAO1; TAO2; TAOK2; Thousand and one amino acid protein 2; thousand and one amino acid protein kinase 2; UNQ2971/PRO7431
Gene Symbols TAOK2
Host Species Rabbit
Immunogen Synthesized peptide derived from human TAOK2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381921, 64666, 9344
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.