Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110054 Supplier Invitrogen™ Supplier No. PA5110054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILNELKPEQV EQLKLIMSKR YDGSQQALDL KGLRSDPDLV AQNIDVVLNR RSCMAATLRI IEENIPELLS LNLSNNRLYR LDDMSSIVQK APNLKILNLS G.

The constitutive transport element (CTE) of type D retroviruses serves as a signal of nuclear export for unspliced viral RNAs. TAP (also known as NXF1) mediates the export of CTE-containing simian type D retroviral RNAs through binding directly to the CTE. TAP is associated with a recognized mRNA export pathway and is a member of the multigene family of NXF proteins. NXF proteins belong to an evolutionarily conserved family of proteins, which are characterized by a leucine-rich-repeat domain (LRR) followed by a region known as the Nuclear Transport Factor 2 (NTF2)-like domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NXF1
Gene Accession No. O88984, Q99JX7, Q9UBU9
Gene Alias Mex 67 homolog; MEX67; Mex67h; modifier of vibrator 1; mRNA export factor TAP; Mvb1; Nuclear RNA export factor 1; nuclear RNA export factor 1 homolog; Nxf1; TAP; tip associated protein; tip associating protein; tip-associated protein; Tip-associating protein; Vdp
Gene Symbols NXF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human NXF1 (NP_0063532).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10482, 53319, 59087
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.