Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAP Tag Monoclonal Antibody (22F12-1E3), DyLight™ 680, Invitrogen™

Catalog No. MA1108D680
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1108D680 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1108D680 Supplier Thermo Scientific Supplier No. MA1108D680

Mouse Monoclonal Antibody

MA1108-D680 detects the Tev cleavage peptide sequence. MA1108-D680 has been successfully used for fluorescent detection of TAP in Western Blot. DyLight 680 has an excitation/emission of 692/712nm.

TAP Tag Monoclonal antibody specifically detects TAP Tag in Tag samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP Tag
Applications Western Blot
Classification Monoclonal
Clone 22F12-1E3
Concentration 1 mg/mL
Conjugate DyLight 680
Formulation PBS with no preservative
Gene Alias Tobacco Etch Virus
Host Species Mouse
Immunogen KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.