Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAP Tag Monoclonal Antibody (22F12-1E3), HRP
GREENER_CHOICE

Catalog No. PIMA1108HRP
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1108HRP 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1108HRP Supplier Invitrogen™ Supplier No. MA1108HRP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1108-HRP detects the Tev cleavage peptide sequence. MA1108-HRP has been successfully used for direct detection of TAP in Western Blot.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Tandem affinity purification (TAP) is an affinity purification method for isolation of TAP-tagged proteins together with associated proteins. The protocol involves the fusion of the 'TAP tag' (typically a calmodulin binding peptide (CBP), a tobacco etch virus protease (TEV protease) cleavage site and Protein A) to the protein of interest. The TAP technique is useful in analyzing in vivo interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP Tag
Applications Western Blot
Classification Monoclonal
Clone 22F12-1E3
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.07% Kathon
Gene Alias Tobacco Etch Virus
Host Species Mouse
Immunogen KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.