Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAP Tag Monoclonal Antibody (22F12-1E3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA1108
Description
MA1-108 detects TAP in Western blot applications. The MA1-108 immunogen is a KLH-conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage. TAP adds about 20 kDa to the size of the protein.
Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Tandem affinity purification (TAP) is an affinity purification method for isolation of TAP-tagged proteins together with associated proteins. The protocol involves the fusion of the TAP tag (typically a calmodulin binding peptide (CBP), a tobacco etch virus protease (TEV protease) cleavage site and Protein A) to the protein of interest. The TAP technique is useful in analyzing in vivo interactions.
Specifications
TAP Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
22F12-1E3 | |
Unconjugated | |
Tobacco Etch Virus | |
KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage. | |
50 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction