Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142408
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142408 Supplier Invitrogen™ Supplier No. PA5142408
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TAP1
Gene Accession No. Q03518
Gene Alias ABC transporter, MHC 1; ABC transporter, MHC, 1; ABC17; ABCB2; ABP-278; ABP-280; ABP-280 homolog; Antigen peptide transporter 1; AOI; APT1; ATP dependent transport protein family member; ATP-binding cassette sub-family B member 2; ATP-binding cassette transporter, major histocompatibility complex, 1; ATP-binding cassette, sub-family B (MDR/TAP), member 2; beta-filamin; Cim; D6S114E; DAAP-57C1.5; FH1; filamin homolog 1; filamin-3; filamin-B; FLJ26666; FLJ41500; FLN1L; FLN-B; Ham1; Ham-1; Histocompatibility antigen modifier 1; LRS1; MTP1; Peptide supply factor 1; peptide transporter involved in antigen processing 1; Peptide transporter PSF1; peptide transporter TAP1; PSF1; PSF-1; Really interesting new gene 4 protein; RING4; SCT; TABP; TAP; tap i; TAP1; Tap-1; TAP1*0102N; TAP1N; Tap2; thyroid autoantigen; transporter 1 ABC (ATP binding cassette); transporter 1 ATP-binding cassette sub-family B; transporter 1, ABC (ATP binding cassette); transporter 1, ATP binding cassette subfamily B member; transporter 1, ATP-binding cassette, sub-family B (MDR/TAP); transporter associated with antigen processing; transporter associated with antigen processing 1; transporter, ABC, MHC, 1; transporter, ATP-binding cassette, major histocompatibility complex, 1; Y3
Gene Symbols TAP1
Host Species Goat
Immunogen Peptide with sequence C-QKFREKLQEIKT (aa 388-399).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6890
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.