Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TAP2, Mouse, Unlabeled, Clone: 429.4,

Catalog No. MABF945 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF945 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF945 Supplier MilliporeSigma™ Supplier No. MABF945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antigen peptide transporter 2 (UniProt Q03519; also known as APT2, ATP-binding cassette sub-family B member 3, Peptide supply factor 2, Peptide transporter involved in antigen processing 2, Peptide transporter PSF2, Peptide transporter TAP2, PSF-2, Really interesting new gene 11 protein) is encoded by the TAP2 (also known as ABCB3, PSF2, RING11, Y1) gene (Gene ID 6891) in human. TAP2 (ABCB3) belongs to the subfamily B of ATP-binding cassette (ABC) transporters, members of which also include TAP1 (ABCB2) and multidrug resistance protein 1 (MDR1, ABCB1, P-gp). TAP is a heterodimeric peptide transporter composed of TAP1 and TAP2 that dimerize via their nucleotide-binding domain (NBD; a.a. 503-510) in a head-to-tail fashion. TAP preferentially mediates the transport of 8- to 16-a.a. peptides into the ER in a process powered by ATP-binding and hydrolysis. TAP1 and TAP2 harbor a specialized N-terminal transmembrane domain (TMD0) that is followed by the TAP half-transporter core module composed of one TMD and one NBD. TMD0 is composed of a 4-TM bundle that serves as the interaction platform for the assembly of the peptide-loading complex (PLC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP2
Applications Western Blot
Classification Monoclonal
Clone 429.4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene TAP2, ABCB3, PSF2, RING11, Y1
Gene Accession No. Q03519
Host Species Mouse
Immunogen Baculovirus-expressed recombinant human TAP2 C-terminal fragment (van Endert, P.M., et al. (1994). Immunity. 1(6):491-500).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000535
Test Specificity Clone 429.4 was subcloned from the original 429.3 hybridoma reported by van Endert, P.M., et al. (1994). Immunity. 1(6):491-500. The epitope lies within TAP2 ABC transporter domain. Clone 429 detected TAP2 in HeLa, but not in the ST-EMO B-lymphoblastoid cell line derived from a patient with homozygous TAP2 deletion (TAP2-/-) (Courtesy of Peter van Endert, Ph.D., Institut Necker Enfants Malades (INEM), Centre De Medecine Moleculaire, Paris, France).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.