Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAP26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570814 Supplier Invitrogen™ Supplier No. PA570814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 85%; Guinea Pig: 91%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 86%; Rat: 100%; Yeast: 82%.

Component of the transcription complexes of the pulmonary surfactant-associated protein-B (SFTPB) and -C (SFTPC). Enhances homeobox protein Nkx-2.1-activated SFTPB and SFTPC promoter activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP26
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC59
Gene Accession No. Q8R2N0
Gene Alias 2300004H16Rik; BR22; Ccdc59; coiled-coil domain containing 59; coiled-coil domain-containing protein 59; D10Ertd718e; HSPC128; RGD1311912; TAP26; Thyroid transcription factor 1-associated protein 26; thyroid transcription factor 1-associated protein 26-like protein; TTF-1-associated protein 26; TTF-1-associated protein BR2; Unknown (protein for IMAGE:8047375)
Gene Symbols CCDC59
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse TAP26.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52713
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.