Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAPA-1 (CD81) Mouse anti-Human, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF206125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2061-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2061-25 Supplier MilliporeSigma Supplier No. MABF206125UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TAPA-1 (CD81), clone 5A6, Cat. No. MABF2061, is a mouse monoclonal antibody that detects TAPA-1 (CD81) and has been tested for use in Flow Cytometry, Inhibition assay, Immunoprecipitation and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAPA-1 (CD81)
Applications Flow Cytometry, Inhibition Assays, Immunoprecipitation, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 containing PBS without azide.
Gene Alias CD81 antigen;26 kDa cell surface protein TAPA-1;Target of the antiproliferative antibody 1;Tetraspanin-28;Tspan-28
Gene Symbols CD81;TAPA1;TSPAN28
Host Species Mouse
Immunogen OCI-LY8 whole cells (Cellosaurus cell line).
Molecular Weight of Antigen 26 kDa calculated.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 5A6 detects TAPA-1 (CD81) in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.