Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tapasin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Tapasin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Tapasin Polyclonal specifically detects Tapasin in Human samples. It is validated for Western Blot.Specifications
Tapasin | |
Polyclonal | |
Rabbit | |
O15533 | |
6892 | |
Synthetic peptides corresponding to TAPBP(TAP binding protein (tapasin)) The peptide sequence was selected from the middle region of TAPBP. Peptide sequence SVSLSGHLQPPPVTTEQHGARYACRIHHPSLPASGRSAEVTLEVAGLSGP. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
NGS17, NGS-17, TAP binding protein (tapasin), tapasin, TAPATAP-associated protein, TPN, TPSNTAP-binding protein | |
TAPBP | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title