Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tapasin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159071
Description
Tapasin Polyclonal specifically detects Tapasin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Tapasin | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
NGS17, NGS-17, TAP binding protein (tapasin), tapasin, TAPATAP-associated protein, TPN, TPSNTAP-binding protein | |
Rabbit | |
Affinity purified | |
RUO | |
6892 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
O15533 | |
TAPBP | |
Synthetic peptides corresponding to TAPBP(TAP binding protein (tapasin)) The peptide sequence was selected from the C terminal of TAPBP. Peptide sequence GEAPPELLCLVSHFYPSGGLEVEWELRGGPGGRSQKAEGQRWLSALRHHS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Bovine: 85%; Pig: 85%; Sheep: 85%. | |
Human, Pig, Bovine, Sheep | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction