Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tapasin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA129450
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA129450 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA129450 Supplier Invitrogen™ Supplier No. PA129450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detects a band of approximately 48 kDa. Positive control of HeLa Cell lysate suggested.

This gene encodes a transmembrane glycoprotein which mediates interaction between newly assembled major histocompatibility complex (MHC) class I molecules and the transporter associated with antigen processing (TAP), which is required for the transport of antigenic peptides across the endoplasmic reticulum membrane. This interaction is essential for optimal peptide loading on the MHC class I molecule. Up to four complexes of MHC class I and this protein may be bound to a single TAP molecule. This protein contains a C-terminal double-lysine motif (KKKAE) known to maintain membrane proteins in the endoplasmic reticulum. This gene lies within the major histocompatibility complex on chromosome 6. Alternative splicing results in three transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tapasin
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide
Gene TAPBP
Gene Accession No. O15533, Q9R233
Gene Alias D17Wsu91e; DADB-159G18.7; NGS17; NGS-17; similar to Tapasin precursor (TPSN) (TPN) (TAP-binding protein) (TAP-associated protein) (NGS-17); TAP binding protein; TAP binding protein (tapasin); TAPA; tapasin; TAP-associated protein; TAP-binding protein; Tapbp; TPN; TPSN
Gene Symbols TAPBP
Host Species Rabbit
Immunogen Synthetic peptide: LGWAAVYLSTCKDSKKKAE, corresponding to amino acids 430-448 of Human Tapasin.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21356, 25217, 504335, 6892
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.