Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAPBPL Polyclonal Antibody
TAPBPL Polyclonal Antibody
Supplier: Thermo Scientific PA558833
Description
TAPBPL Polyclonal Antibody for Western Blot, IHC

Specifications
TAPBPL | |
Polyclonal | |
Unconjugated | |
TAPBPL | |
BC017613; TAP binding protein like; TAP binding protein related; TAP binding protein-like; tapasin-like; TAPASIN-R; tapasin-related protein; tapasin-related protein-like; TAP-binding protein-like; TAP-binding protein-related protein; Tapbpl; Tapbplr; TAPBPL-R; TAPBPR; TAPBP-R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55080 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BX59 | |
TAPBPL | |
Recombinant protein corresponding to Human TAPBPL. Recombinant protein control fragment (Product #RP-90292). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction