Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Taped Tissue Cassette, 45° angle front
SDP

Catalog No. p-200025600
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Tissue cassettes, taped for automation, hinged lid separate, 45° angle front, taped in stacks of 40, 50 stacks/case.

These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage. Molded from acetal, they keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.

This model differs by the special hinge that holds the base and cover together. The hinge design configuration allows for multiple opening and closing of the lid.

Features:

  • Made of acetal
  • Anterior writing area at a 45° angle
  • Easy-to-assemble disposable cover with special hinge
  • Large tab for convenient and easy opening
  • Covers packaged separately

Specifications

Closure Included Yes
Closure Type Lid
Color Aqua
Product Type Tissue Cassette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.