Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566719 Supplier Invitrogen™ Supplier No. PA566719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITVPKQSDSQP NSDNLSRHGE CGKKQVSYRT DIVGGVPIIT PTQKEEVNEC GESIDRNNLK RSQSHLPYFT PKPPQ Highest antigen sequence identity to the following orthologs - mouse 91%, rat 92%.

Phosphatidylinositol-4,5-bisphosphate 3-phosphatase adapter protein 1 (TAPP1) is a gene located on chromosome 1q25.2, encoding a protein involved in intracellular signaling pathways that regulate cellular processes such as proliferation, survival, and migration. TAPP1 is characterized by containing pleckstrin homology (PH) domains, which allow it to bind specifically to phosphoinositide lipids, particularly phosphatidylinositol (3,4) bisphosphate. This interaction is crucial for the localization of TAPP1 to cell membranes where it can act as an adaptor protein, modulating the activity of downstream signaling molecules and pathways. TAPP1 is expressed in various tissues and plays a role in immune cell function, influencing processes like cell motility and cytoskeletal dynamics. Research into TAPP1 focuses on its role in immune regulation and signaling, with implications for understanding its function in disease contexts such as cancer and autoimmune disorders where phosphoinositide signaling is disrupted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAPP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHA1
Gene Accession No. Q9HB21
Gene Alias AA960558; C920009D07Rik; PH domain-containing family A member 1; pleckstrin homology domain containing A1; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1; pleckstrin homology domain-containing family A member 1; pleckstrin homology domain-containing protein family A member 1; PLEKHA1; RGD1564153; tandem PH domain containing protein-1; tandem PH domain-containing protein 1; TAPP1; TAPP-1
Gene Symbols PLEKHA1
Host Species Rabbit
Immunogen Recombinant Human TAPP1. Recombinant protein control fragment (Product #RP-107371).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.