Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAPP1 Recombinant Rabbit Monoclonal Antibody (16C9)
GREENER_CHOICE

Catalog No. FEMA550061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550061 Supplier Thermo Scientific Supplier No. MA550061
Add to Cart
Add to Cart

TAPP1 Recombinant Rabbit Monoclonal Antibody (16C9)

TAPP1 Recombinant Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TAPP1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 16C9
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLEKHA1
Gene Accession No. Q9HB21
Gene Alias AA960558; C920009D07Rik; PH domain-containing family A member 1; pleckstrin homology domain containing A1; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1; pleckstrin homology domain-containing family A member 1; pleckstrin homology domain-containing protein family A member 1; PLEKHA1; RGD1564153; tandem PH domain containing protein-1; tandem PH domain-containing protein 1; TAPP1; TAPP-1
Gene Symbols PLEKHA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLEKHA1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59338
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.